Learn More
Thermo Scientific™ PT100 External Probe
Catalog No. CIC0001447
Change view
Click to view available options
Type:
Probe
Catalog No. Type
CIC0001447 Probe
1 options
Catalog No. CIC0001447 Supplier Thermo Scientific™ Supplier No. CIC0001447
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
  • Composed of AISI 316Ti, a titanium-stabilized version of the 316 stainless steel
  • AISI 316Ti is a type of austenitic chromium-nickel stainless steel specifically designed to resist intergranular corrosion at high temperatures
Specifications
Description PT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
For Use With (Equipment) Cimarec and Supernuova Hotplates and Hotplate Stirrers
Temperature (Metric) 0°C to 400°C
Type Probe
Display Digital
Material Stainless Steel