missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermco™ Pt100 Digital Dual Probe Reference Thermometer
Description
Requires
Additional purchase of probe(s); additional purchase of calibration required
Features:
- 2-channel resistance thermometer
- Automatically detects Thermco Pt100 smartprobes wich have an integrated Eprom where the calibration characteristic of each probe is transferred automatically to the instrument
- High performance making it suitable as primary reference standard
- Recording maximum, minimum, hold and average values
- High measuring accuracy
- Selectable reading function: 1 second to 30 minutes
- Calibration coefficients (R0ABC) will automatically transfer from the probe to the instrument
Specifications
Specifications
| Dimensions (L x W x H) | 200 mm x 85 mm x 40 mm |
| Display Dimensions | 44 mm x 70 mm |
| Disposable | Reusable |
| Height (Metric) | 40 mm |
| Includes | Mounting devise for probe handle (probe not included); USB cable |
| Length (Metric) | 200 mm |
| Measurement Time | 1 sec. to 30 min. |
| Probe Type | Dual channel |
| Resolution | ±0.001°C between -50°C to 199.99°C; 0.025% reading remaining range |
| Weight (Metric) | 300 g |
| Show More |
Do not dispose of electronic devises or batteries as household waste. Follow appropriate local, state and federal regulations.