missing translation for 'onlineSavingsMsg'
Learn More

Psoralen, 97%, Thermo Scientific Chemicals

Catalog No. AAJ60042MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60042MC 100 mg
AAJ60042ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60042MC Supplier Thermo Scientific Chemicals Supplier No. J60042MC

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66-97-7
Molecular Formula C11H6O3
Molecular Weight (g/mol) 186.17
MDL Number MFCD00010520
InChI Key ZCCUUQDIBDJBTK-UHFFFAOYSA-N
Synonym psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
PubChem CID 6199
ChEBI CHEBI:27616
IUPAC Name 7H-furo[3,2-g]chromen-7-one
SMILES O=C1OC2=CC3=C(C=CO3)C=C2C=C1

Specifications

Melting Point 163°C to 164°C
Color White
Quantity 100 mg
Beilstein 152784
Merck Index 14,7928
Formula Weight 186.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Psoralen
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-639-7
RTECSNumber LV0944000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only