missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PSMA/FOLH1/NAALADase I Recombinant Protein Antigen

Catalog No. NB036511
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036511 Supplier R&D Systems™ Supplier No. NBP288924PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results

Specifications

Gene ID (Entrez) 2346
Product Line Novus
Protein Family PSMA/FOLH1/NAALADase I
Purity >80%, by SDS-PAGE
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS, 1 M Urea, pH 7.4.
Gene Symbol FOLH1
Quantity 100 μg
Research Category Cancer, Prostate Cancer
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55081. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml