Learn More
New England Biolab Ltd PshAI 5000 units
Catalog No. 50812307
Encompass
A restriction endonuclease that recognizes the sequence GACNN_^NNGTC.
Catalog No. 50812307 Supplier New England Biolab Ltd Supplier No. R0593L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A restriction endonuclease that recognizes the sequence GACNN_^NNGTC.