Learn More
Abcam Inc PSB 11 hydrochloride, competitive human A3 receptor antagonist, 50MG
Catalog No. 502707049
Encompass
MW 331.8 Da, Purity >99%. Potent, selective, competitive human A3 receptor antagonist (Ki values are 2.3 nM and 10 M for human and rat A3 receptors, respectively), Antagonises human A1 and A2A receptors. Shows inverse agonist effects in the [35S]GTPS binding assay. Shows anti-inflammatory effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502707049 Supplier Abcam Inc Supplier No. AB12099550MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 331.8 Da, Purity >99%. Potent, selective, competitive human A3 receptor antagonist (Ki values are 2.3 nM and 10 M for human and rat A3 receptors, respectively), Antagonises human A1 and A2A receptors. Shows inverse agonist effects in the [35S]GTPS binding assay. Shows anti-inflammatory effects.
The product is subject to the following: Abcam Restricted Use Statement