missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate Dibasic, Anhydrous, Powder, 98%, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Crystalline Powder or Needles |
| CAS Max % | 1 |
| Quantity | 100 lb. |
| Packaging | Poly Bottle |
| Molecular Weight (g/mol) | 136.06 |
| Percent Purity | 98% |
| Grade | Reagent |
| Chemical Name or Material | Calcium Phosphate Dibasic, Anhydrous, Powder |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.