missing translation for 'onlineSavingsMsg'
Learn More

High-Tech Conversions ProWorks™ High Dexterity Nitrile Gloves

Catalog No. 17444091
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17444091 Small
17444090 Medium
17444089 Large
17444092 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17444091 Supplier High-Tech Conversions Supplier No. GLN103FS
Only null left

ProWorks™ High Dexterity Nitrile Gloves are powder free nitrile gloves that offer high dexterity and are suitable for industrial use.

  • Chemical resistant
  • Textured fingers
  • Meets FDA 21 CFR for food contact
  • 100/box, 10 boxes/case

Specifications

Certifications/Compliance FDA 21 CFR
Chlorinated No
Accelerator Free No
Applications Industrial, Laboratory
AQL 2.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 3 mil
Length (English) 9.5 in.
Show More Show Less