Learn More
New England Biolab Ltd ProtoScript First Strand cDNA Synthesis Kit 150 reactions
Catalog No. NC9004281
Encompass
ProtoScript First Strand cDNA Synthesis Kit features two optimized mixes, M-MuLV Enzyme Mix and M-MuLV Reaction Mix. M-MuLV Enzyme Mix combines M-MuLV Reverse Transcriptase and Murine RNase Inhibitor, while M-MuLV Reaction Mix contains dNTPs and an optimized buffer. The kit also contains two optimized primers for reverse transcription and nuclease-free water. An anchored oligo-dT primer [d(T)23VN] forces the primer to anneal to the beginning of the polyA tail. The optimized Random Primer Mix provides random and consistent priming sites covering the entire RNA template including both mRNAs and non-polyadenylated RNAs. The first strand cDNA product generated is more than 10 kb.
Catalog No. NC9004281 Supplier New England Biolab Ltd Supplier No. E6300L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ProtoScript First Strand cDNA Synthesis Kit features two optimized mixes, M-MuLV Enzyme Mix and M-MuLV Reaction Mix. M-MuLV Enzyme Mix combines M-MuLV Reverse Transcriptase and Murine RNase Inhibitor, while M-MuLV Reaction Mix contains dNTPs and an optimized buffer. The kit also contains two optimized primers for reverse transcription and nuclease-free water. An anchored oligo-dT primer [d(T)23VN] forces the primer to anneal to the beginning of the polyA tail. The optimized Random Primer Mix provides random and consistent priming sites covering the entire RNA template including both mRNAs and non-polyadenylated RNAs. The first strand cDNA product generated is more than 10 kb.