missing translation for 'onlineSavingsMsg'
Learn More

Synbiosis™ Protos 3
GREENER_CHOICE

Catalog No. 01257326
Change view
Click to view available options
Display Type:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
01257326 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257326 Supplier Synbiosis™ Supplier No. PROTOS3

missing translation for 'validMainframeMsg'

The Protos 3 is an automated colony counter and chromogenic identification system. Using its sensitive CCD camera and unique lighting coupled with powerful analysis software, Protos 3 counts colonies in seconds and automatically identifies microbial species by their color on chromogenic plates.

  • Fully traceable, reproducible results
    • Can input plate identification using 1D or 2D barcode
    • Automatic capability to transfer and print results and images
    • Saves counts to Excel to eliminate keying errors
    • Saves images in TIFF, Excel, and report formats
    • Archives results in accessible SQL database
  • Increases throughput
    • Accurately detects colonies as small as 0.043 mm
    • Precisely counts individual colonies in seconds
    • Capable of analyzing 75 plates in 5 minutes
  • Excellent detection performance
    • True color recognition using unique 3 color lighting
    • Identifies multiple different colony colors on one plate
    • Analyzes all plate colors with a choice of light and dark backgrounds
    • Classifies by size, color, and shape
  • Simple to use
    • Intuitive software
    • Easily connects to computer via USB
    • Corrects for agar defects, touching colonies, or sample debris
    • Marks colonies with symbols to show each counted
    • Easy to exclude areas from a count
    • Adjustable counting area
  • Multiple plate reading functions
    • Pour plates
    • Spiral plates
    • Multi-sector plates
    • Gridded plates
    • Streak plates
    • Dilution series plates
    • Species identification
    • Large plates (up to 150 mm)

Specifications

Product Type Automated Colony Counter and Chromogenic Identification System
Display Type None
Memory Injection-molded Resin
Product Line Synbiosis
Sample Size 30 to 150 mm
Voltage 115 V
For Use With (Application) Automatic colony counting and chromogenic identification