missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ SafeClear 2 Xylene Substitute, Fisherbrand™

Catalog No. 11002233
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002233 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002233 Supplier Statlab Supplier No. 11002233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

Aliphatic hydrocarbon for processing and staining.

  • A safer clearing agent without the hazards of xylene.
  • Low odor
  • Flash point 145°F

Specifications

Product Type Xylene Substitute
Quantity 1 gal.
Form Solution

Made in the USA