missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Parasitology Two-Vial Kits, Modified Cu-PVA/10% Formalin Kits

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.

$133.07

Specifications

Product Type Parasitology Two Vial Kit
Quantity 2 Pk, 10 Pk./Cs.
Volume (English) 1.01 oz.
Volume (Metric) 30 mL
View More Specs

Products 1
Catalog Number Mfr. No. Media Type Price Quantity & Availability  
Catalog Number Mfr. No. Media Type Price Quantity & Availability  
2300545
View Documents Promotion Details
Fisher Healthcare™
2300545
Modified (Cu) PVA, Formalin
Pack of 20 Each for $133.07
Only null left
 
Description

Description

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Two-Vial Kits.

Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA):

  • Mercury-free fixative is for use as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin

Sodium Acetate Acetic Acid Formalin (SAF):

  • Mercury-free preservative/fixative is ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • Slides fixed with SAF may be stained using trichrome or hematoxylin

10% Buffered Formalin:

  • All-purpose fixative is appropriate for helminth eggs and larvae and for protozoan cysts
  • Buffering system helps maintain organism morphology
  • Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts

Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA):

  • Fixitive acts as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin

Culture & Sensitivity (C & S Media):

  • C&S is for collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and CholeraVibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours

Clean Vials:

  • Empty 30mL vials are for transporting specimens

Ordering Alerts:

Supplied as 10 packs of two vials each

Note:

All PROTOCOL Parasitology products are mercury free.

Specifications

Specifications

Parasitology Two Vial Kit
2 Pk, 10 Pk./Cs.
1.01 oz.
30 mL
Videos
SDS
Documents

Documents