Learn More
Fisher Healthcare™ PROTOCOL™ Parasitology Single-Vial Kits, Clean Vials
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.
Supplier: Fisher Healthcare™ 23 005 43
Description
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Single-Vial Kits.
Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)
- Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
- Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin
Sodium Acetate Acetic Acid Formalin (SAF)
- Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
- May be stained using trichrome or hematoxylin
10% Neutral Buffered Formalin
- All-purpose fixative appropriate for helminth eggs and larvae, and for protozoan cysts
- Buffering system helps to maintain organism morphology
- Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts
Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)
- Acts as a preservative/fixative for protozoan trophozoites
- Slides fixed with Modified PVA may be stained using trichrome or hematoxylin
Culture and Sensitivity (C & S Media)
- For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio
- Bacterial population is maintained for optimum recovery, up to 96 hours
Cary-Blair Media
- Maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio for up to 96 hours
Clean Vials
- Empty, 30mL vials for transporting specimens
Ordering Alerts:
Supplied as pack of 20 vials
Note:
All PROTOCOL Parasitology products are mercury free.
Chemical Identifiers
| 1.01 oz. | |
| 30 mL |
Specifications
| Parasitology Single Vial Kit | |
| 1 Vial/Kit, 20 Vials/Pk. | |
| 30 mL |
| C&S | |
| 1.01 oz. |