missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Parasitology Single-Vial Kits, Clean Vials

Catalog No. 2300541
Change view
Click to view available options
Media Type:
10% Buffered Formalin
C&S
Cary-Blair
SAF
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
2300541 SAF
2300546 10% Buffered Formalin
2300543 C&S
2300547 Cary-Blair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2300541 Supplier Fisher Healthcare™ Supplier No. J2300541
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Single-Vial Kits.

Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)

  • Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin

Sodium Acetate Acetic Acid Formalin (SAF)

  • Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • May be stained using trichrome or hematoxylin

10% Neutral Buffered Formalin

  • All-purpose fixative appropriate for helminth eggs and larvae, and for protozoan cysts
  • Buffering system helps to maintain organism morphology
  • Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts

Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)

  • Acts as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin

Culture and Sensitivity (C & S Media)

  • For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours

Cary-Blair Media

  • Maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio for up to 96 hours

Clean Vials

  • Empty, 30mL vials for transporting specimens

Ordering Alerts:

Supplied as pack of 20 vials

Note:

All PROTOCOL Parasitology products are mercury free.

Specifications

Product Type Parasitology Single Vial Kit
Media Type SAF
Quantity 20 Vials/Pk.
Volume (English) 1.01 oz.
Volume (Metric) 30 mL