missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Healthcare™ PROTOCOL™ Parasitology Concentrate System
Description
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Concentrate Systems.
50mL Concentrate System
- Complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens
- The concentration system creates a completely closed system that minimizes exposure to infectious agents and fits with any of the PROTOCOL System vials
- Each 50mL Concentrate System Kit contains: 120 Filtering Devices, 120 Conical Centrifuge Tubes, 120 Screw Caps and 30mL Surfactant
- Specimens collected and transported in 10% formalin, SAF or Clean vials may be used with this concentration system
15mL Concentrate System
- Complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens
- Each 15mL Concentrate System Kit contains: 50 Filtering Devices, 50 Disposable 15mL centrifuge tubes and caps, Reagent A (Surfactant) and Reagent B (Ethyl Acetate)
- Specimens collected and transported in 10% formalin, SAF or Clean vials may be used with this concentration system
Note:
All PROTOCOL Parasitology products are mercury free.
Specifications
Specifications
| Product Type | Concentrate System |
| Description | Concentrate System - A complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens. |
| Includes | 120 Filtering Devices, 120 Conical Centrifuge Tubes, 120 Screw Caps, 30 mL Surfactant |
| Quantity | 120/Pk. |
| Volume (English) | 1.69 oz. |
| Volume (Metric) | 50 mL |