missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Parasitology Ancillary Products, Trichrome Stain

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.

Supplier:  Fisher Healthcare™ 23 005 40

 View more versions of this product

Catalog No. 2300540


Only null left

Description

Description

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Ancillary Products.

Trichrome Stain

  • Rapid staining procedure producing uniformly stained smears of intestinal protozoa, yeast and human cells
  • Provides excellent contrast facilitating the detection of intestinal parasites while differentiating organisms from fecal debris and artifacts
  • Supplied in a 500mL bottle

Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA) Bulk

  • Mercury-free fixative for protozoan trophozoites in a 500mL bottle

Ethyl Acetate

  • Supplied in a 225mL bottle

Note:

All PROTOCOL Parasitology products are mercury free.

Specifications

Specifications

Zinc PVA Bulk
Zn-PVA Bulk - A mercury-free fixative for use as a preservative/fixative for protozoanm trophozoites.
16.91 oz.
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions