missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ Mounting Media, Fisherbrand™

Catalog No. 11002231
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002231 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002231 Supplier Statlab Supplier No. 11002231

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

A rapid-drying, xylene-based mounting medium.

  • For mounting and preserving microscope slide specimens
  • Acts as a protective layer between the tissue and the coverslip while maintaining a suitable refractive index for clear imaging.

Specifications

Quantity 16 oz.
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 473 mL

Made in the USA