missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ Histosolv-X Xylene Substitute, Fisherbrand™

Catalog No. 11002213
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002213 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002213 Supplier Statlab Supplier No. 11002213

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

100% d-Limonene for processing and staining.

  • Xylene Substitute featuring a mild citrus odor with exceptionally high purity.
  • A xylene alternative clearing agent
  • Flash point 122 degrees F.

Specifications

Product Type Xylene Substitute
Quantity 1 gal.
Form Solution

Made in the USA