missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ Formalin, Fisherbrand™

Catalog No. p-200109698
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2.5 gal.
5 gal.
55 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002205 1 gal.
11002206 2.5 gal.
11002207 5 gal.
11002208 55 gal.
11002229 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002205 Supplier Statlab Supplier No. 11002205
Only null left
Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use buffered formalin in bulk packaging.

  • 10% neutral buffered formalin fixative
  • Used to preserve the cellular structure and morphology of tissue for examination under a microscope by preventing decay and maintaining its molecular integrity during the tissue processing steps for staining and analysis
  • Featuring a pH level of 7.0

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 1 gal.
Solution Type Formalin
Form Solution

Made in the USA