missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ Formalin, Fisherbrand™

Catalog No. 11002207
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2.5 gal.
5 gal.
55 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002207 5 gal.
11002205 1 gal.
11002206 2.5 gal.
11002208 55 gal.
11002229 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002207 Supplier Statlab Supplier No. 11002207
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use buffered formalin in bulk packaging.

  • 10% neutral buffered formalin fixative
  • Used to preserve the cellular structure and morphology of tissue for examination under a microscope by preventing decay and maintaining its molecular integrity during the tissue processing steps for staining and analysis
  • Featuring a pH level of 7.0

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 5 gal.
Solution Type Formalin
Form Solution

Made in the USA