missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ Formalin, Fisherbrand™

Catalog No. 11002210
Change view
Click to view available options
Quantity:
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002210 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002210 Supplier Statlab Supplier No. 11002210

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

20% NBF intended as a general fixative for all tissues to be examined in light microscopy.

  • Solution is often used for surgical and necropsy specimens, and is also excellent for brain tissue.
  • A general purpose fixative for light microscopy analysis

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 5 gal.
Solution Type Formalin
Form Solution

Made in the USA