Learn More
Protocol™ Decalcifying Solution A, Fisherbrand™
A gentle decalcifier formulated from semiconductor grade hydrochloric acid, EDTA, and highly purified reagent grade water.
$75.25 - $188.57
Specifications
| Product Type | Decalcifying Solution |
|---|---|
| Solution Type | Decalcifier |
| Form | Solution |
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Description
- Can be used to safely decalcify all types of tissue samples from needle biopsies to cortical bone
- Mild enough that most samples can be left in overnight
Specifications
| Decalcifying Solution | |
| Decalcifier | |
| Solution |
Made in the USA