missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocol™ Carson's Millonig Formalin, Fisherbrand™
Description
- Formula can be used in place of traditional 10% NBF.
- A dual-purpose fixative that can be used for both light and electron microscopy.
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Histological |
| Product Type | Formalin |
| Quantity | 5 gal. |
| Solution Type | Formalin |
| Form | Solution |
Made in the USA