missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ProteoExtract™ Protein Precipitation Kit

Catalog No. 5391801KIT
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5391801KIT 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5391801KIT Supplier MilliporeSigma™ Supplier No. 5391801KIT
Only null left

Yields protein samples with significantly lower conductivity than other precipitation methods

  • Convenient and highly reproducible kit for precipitation and clean-up of proteins
  • Yield of precipitated proteins is higher than that of traditional methods with improved solubility of precipitated protein
  • Contains sufficient components to precipitate 200 samples of ≤200μLl
  • Requires 150mL of ethanol (not provided)
  • Kit delivers protein solutions of very low conductivity

Specifications

Quantity 1 kit
Format 200 Precipitations
Kit Contents Precipitant 1, Precipitant 2, Precipitant 3, Precipitant 4, Wash Solution, and a user protocol
Content And Storage Store at+15°C to+30°C