The ProteinWorks Auto-eXpress Low 3 Digest Kit simplifies and accelerates the digestion of immunopurified plasma & serum samples for LC-MS protein quantification via the surrogate peptide approach. The kit includes a robust digest protocol and pre-measured, lot-traceable reagents in an automation friendly format for the denaturation, digestion and quench of proteins in biological matrices. This product is suitable for high protein content samples (0.2 - 1.0 mg total protein) and contains the following: ProtienWorks Low Digest Ambient Kit and ProteinWorks Low Digest Trypsin Kit