Learn More
Acrobiosystems ACROBiosystems Recombinant Protein L Protein, His Tag DFS Item
Supplier:  ACROBiosystems Inc RPLP31411G
Recombinant Protein;1G;Recombinant Protein L, His Tag, also known as Recombinant Peptostreptococcus magnus Protein L, was expressed in E.Coli at ACRObiosystems. ;Recombinant Protein L, His Tag is fused with the polyhistidine tag at N-terminus and a single cysteine at C-terminus. The reducing (R) protein migrates as a 36-38 kDa polypeptide .Protein L Protein can bind to VL-Kappa.
Catalog No. 50-195-6641
Only null left
Explore available promotions