missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies Protein Synthesis Manipulative Kit

Catalog No. S65998
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S65998 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65998 Supplier United Scientific Supplies Supplier No. PSYKIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Teach protein synthesis with hands-on manipulatives.

The kit was developed by the same teacher who brought you recombinant DNA technology manipulatives. Now available are manipulatives for teaching protein synthesis. Whether you have taught this content a thousand times or this is your first, you will find that all students can learn this material. What makes this kit different from any other protein synthesis instructional materials:

  • 33 Teacher Manipulatives: These colorful, large DNA, mRNA, ribosome, tRNA and amino acid models attach to your blackboard and can be seen from the back of the classroom. You simulate the process for your students at your own pace, allowing students to ask questions as you proceed.
  • 180 Student Manipulatives: Students work at their tables using smaller size models to work through the process and internalize key concepts. Includes 5 sets of student materials, sufficient for a class of 30 students. Assessment: Each student is given a unique DNA sequence and is asked to identify the resulting amino acid sequence. Verification of the sequence is a snap using the included teacher key.
  • No Consumables: The kit can be used over and over again, and can be shared by the entire science department.
  • Downloadable Instructions/Activity Guide: A link to a file demonstrating how to use the kit is included. Students see protein synthesis in action, model it and are then assessed on the lessons. It is a complete package that makes complex biological processes fun for students and easy to teach.

Order Info

Class size: 30 students working in groups of five.

Specifications

Grade 5 to 12
For Use With (Application) For teaching protein synthesis
Includes Teacher manipulatives, student manipulatives and assessments, instructional cd-rom with suggested activities
Class Size 30 Students
Product Type Protein Synthesis Manipulatives Kit