Learn More
Eppendorf™ Protein LoBind™ Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
Supplier: Eppendorf™ 0030108450
Description
- Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
- Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
- Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
- Snap, screw, and SnapTec™ cap options available.
Chemical Identifiers
| HDPE | |
| Non-sterile | |
| Microcentrifuge Tube (PCR Clean, Protein LoBind) |
| Polypropylene | |
| Conical | |
| PCR Clean (Non-sterile) |
Specifications
| HDPE | |
| 2 mL | |
| 500 Pk. | |
| Non-sterile | |
| Conical | |
| Microcentrifuge Tube (PCR Clean, Protein LoBind) | |
| PCR Clean (Non-sterile) |
| Colorless | |
| Polypropylene | |
| 1 Bag x 500 Tubes | |
| Autoclavable | |
| Snap Cap | |
| 18,000 x g |