missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Deepwell Plates

Catalog No. 05414230
Change view
Click to view available options
Color:
White
White, Clear Bottom
Yellow
No. of Wells:
384
96
Volume (Metric) Well:
140 μL
200 μL
500 μL
1000 μL
2000 μL
Packaging:
10 Bags x 12 Plates
10 Bags x 24 Plates
10 Bags x 8 Plates
5 Bags x 4 Plates
5 Bags x 8 Plates
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells Volume (Metric) Well Packaging
05414230 Yellow 96 500 μL 5 Bags x 8 Plates
E951032107 White 96 500 μL 5 Bags x 8 Plates
05414233 White 96 500 μL 10 Bags x 12 Plates
E951032905 White 96 1000 μL 5 Bags x 4 Plates
05414239 Yellow 96 1000 μL 5 Bags x 4 Plates
E951033308 White, Clear Bottom 96 1000 μL 10 Bags x 8 Plates
E0030504305 White 96 2000 μL 5 Bags x 4 Plates
E951040589 White 384 140 μL 10 Bags x 8 Plates
05414247 White 384 140 μL 10 Bags x 24 Plates
E951031305 White 384 200 μL 5 Bags x 8 Plates
05414222 White 384 200 μL 10 Bags x 12 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 05414230 Supplier Eppendorf™ Supplier No. 951032123
Only null left

Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.

  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.

Specifications

Autoclavable Autoclavable
Packaging 5 Bags x 8 Plates
Color Yellow
Product Type Protein LoBind™ Plate
Material Polypropylene
Volume (Metric) Well 500 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Preparation or Storage of Protein, Peptide or Antibody Samples
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 40 Pk.
Well Color Colorless
Dimensions (L x W x H) 85.5 x 127.8 x 27.1 mm
Show More Show Less