missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Protein LoBind™ Deepwell Plates
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing in plates.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Packaging | 10 Bags x 12 Plates |
| Color | White |
| Product Type | Protein LoBind™ Plate |
| Material | Polypropylene |
| Volume (Metric) Well | 200 μL |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| For Use With (Application) | Preparation or Storage of Protein, Peptide or Antibody Samples |
| Max. RCF | 6000 x g |
| Show More |