missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protein Loading Buffer for Native-PAGE, 5X

Catalog No. ICN08W00016
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN08W00016 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN08W00016 Supplier MP Biomedicals™ Supplier No. 08W00016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5X Protein Loading Buffer for Native-PAGE can be used to prepare protein samples for non-denaturing gel electrophoresis using native PAGE gels.

  • Modified, 5X concentrated protein loading buffer containing bromophenol blue dye
  • 5X protein loading dye for Native-PAGE electrophoresis
  • Contains bromophenol blue dye

Specifications

Buffer Protein Loading
Color Blue
Concentration 5X
Detection Method Electrophoresis
Gel Type Native-PAGE
Volume (Metric) 1 mL
Form Liquid
Quantity 1 Vial
Storage Requirements -20°C
Product Type Loading Buffer

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.