Learn More
Protein A HRP Conjugate , TCI America™
Labeled with HRP
Supplier: TCI America P24661VIAL
Description
Protein A is a type I membrane protein produced by several strains of Staphylococcus aureus. It has high-affinity binding sites that enable the purification of IgG obtained from various species such as humans, rabbit, mouse, and bovine. Recombinant Protein A is produced in E. coli and essentially functions in the same way as native Protein A.Specifications
| Protein A | |
| 0.2 mg | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. |
| HRP | |
| Liquid |