missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein A–FITC Conjugate

Catalog No. 101011
Encompass
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
101011 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 101011 Supplier Invitrogen™ Supplier No. 101011
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein A–FITC Conjugate

Protein A (from Staphylococcus aureus, Cowan I strain) is conjugated with fluorescein isothiocyanate (FITC) and is purified by molecular sieving to remove the unbound FITC and unconjugated Protein A. The product is in 10 mM phosphate buffered saline, pH 7.4, containing 1% ovalbumin, 40% glycerol, and 0.1% sodium azide.

Specifications

Conjugate Fluorescein
Quantity 2 mL
Form Solution
Protein Subtype Protein A
Content And Storage Store at 2-8°C
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.