Learn More
TCI America™ Protein A Biotin Conjugate
Labeled with Biotin
Supplier:  TCI America™ P24071VIAL
Catalog No. P2407VIAL
Only null left
Explore available promotions
Description
Description
Protein A is a type I membrane protein produced by several strains of Staphylococcus aureus. It has high-affinity binding sites that enable the purification of IgG obtained from various species such as humans, rabbit, mouse, and bovine. Recombinant Protein A is produced in E. coli and essentially functions in the same way as native Protein A.
Specifications
Specifications
Protein A
1 mg
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
Biotin
Liquid
Product Suggestions
Product Suggestions