missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ protein A, Alexa Fluor™ 546 conjugate

Catalog No. P11049
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11049 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11049 Supplier Invitrogen™ Supplier No. P11049
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

protein A, Alexa Fluor™ 546 conjugate

Our Alexa Fluor 546 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, orange-fluorescent probe (absorption/emission maxima ∼556/573 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.

Specifications

Conjugate Alexa Fluor 546
Quantity 1 mg
Product Line Alexa Fluor
Form Lyophilized Powder
Protein Subtype Protein A
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.