missing translation for 'onlineSavingsMsg'
Learn More

DenLine™ Uniforms Protection Plus™ DL171 Unisex Mid-Length Lab Coats

Catalog No. p-4381737
Change view
Click to view available options
Color:
Black
Size:
2X-Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22412203 Black 2X-Large
22412204 Black 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22412203 Supplier DenLine™ Uniforms Supplier No. 6040805
Only null left

Mid-length lab coats provide AAMI PB70 Level 2 fluid resistance against incidental splash of blood, body fluids, aerosols, and all aqueous liquids. Suitable for cleanroom use.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • High barrier protection against strike through from incidental splash of blood
  • Hydrostatic Pressure >32.5 CM (AATCC 127) and Impact Penetration <1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles.
  • Complies with OSHA 29 CFR 1910
  • 360 degree protection front, shoulders, and back
  • Fluid resistant fabric, AAMI PB70 Level 2 Barrier
  • No cotton fabric used on shoulders or back as it would not meet OSHA 29 if splashed with blood or OPIM traveling rearward either side of the employee's head
  • Mid-Length 34 in. coat provides wearers 5 ft. 4 in. and shorter frontal coverage to knees
  • Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
  • High air permeability (35 CFM) is used to make the back of coat for wearer comfort
  • Overall air permeability of coat front and back is 18 CFM (ASTM D737)
  • Stain fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Hang loop outside back of collar
  • Side vents for pant pocket access
  • Choice of 5 Colors White, Ceil Blue, Royal, Black and Green, Sizes XS to 3XL

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polyester, Carbon
Color Black
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static, Fluids
Style Mid-length
No. of Pockets 3
Description Durable Knit Cuffs for Overgloving, Bleach Safe Colors
Collar Type Traditional
Latex Free Yes
Size (English) Chest 50 to 52 in.
Size (English) Hip 46 to 49.5 in.
Size (Metric) Chest 127 to 132 cm
Size (Metric) Hip 117 to 126 cm
Certifications/Compliance OSHA, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanroom ISO 5/Class 100
Length (English) 34 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 42.5 in.
Length (Metric) 86 cm
Length (Metric) Cuff 10 cm
Autoclavable Autoclavable
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less