missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protection Filter Set

Catalog No. 361046341
Change view
Click to view available options
Volume (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
361046341 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361046341 Supplier Eppendorf™ Supplier No. 4920623005
Only null left

Designed for use with Eppendorf Reference™ 2, Eppendorf Research™ plus and Eppendorf Xplorer™ (plus) pipettes

  • Available in a variety of colors
  • Protection Filter Sets include 10 filters and 1 filter sleeve
  • For fixed- and variable-volume pipettes

Specifications

Color Violet
Type Protection Filter
Volume (Metric) 5 mL
For Use With (Equipment) 0.5 to 5mL variable Eppendorf Reference™ 2, Eppendorf Research plus and Eppendorf Xplorer™ (plus)
Includes 10 filters with 1 filter sleeve
Quantity 1 Each