Learn More
Merck Emd Millipore Protease from Streptomyces griseus powder, BioReagent, suitable for mouse embryo cell culture, >=3.5 units/mg solid | 9036-06-0 | MFCD00132092 | 1G
Catalog No. NC1387413
Encompass
Protease from Streptomyces griseus powder, BioReagent, suitable for mouse embryo cell culture, >=3.5 units/mg solid | 9036-06-0 | MFCD00132092 | 1G
Catalog No. NC1387413 Supplier Merck Emd Millipore Supplier No. P88111G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protease from Streptomyces griseus powder, BioReagent, suitable for mouse embryo cell culture, >=3.5 units/mg solid | 9036-06-0 | MFCD00132092 | 1G