Learn More
Enzo Life Sciences Proteasome 20S beta1 subunit monoclonal antibody (MCP421) (100 l) DFS Item
Supplier:  Enzo BMLPW81400100
Clone: MCP421. Host: Mouse. Isotype: IgG1. Immunogen: Dinitrophenylated proteasomes. UniProt: P28072. Source: Purified from mouse ascites. Species Reactivity: Human, Yeast, Rabbit. Specificity: Recognizes the 1 subunit of the 20S proteasome. Applications: IHC, WB. Purity: Protein G-affinity purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20C.
Catalog No. NC1528209
Only null left
Explore available promotions