missing translation for 'onlineSavingsMsg'
Learn More

Therapak™ Prostate Biopsy Collection and Transport Kit

Complete with your choice of 6, 8, 12, 14 or 16 prefilled jars with 10mL formalin solution

Supplier:  Therapak™ 38869

 View more versions of this product

Catalog No. 22-131-457


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Biopsy Collection and Transport Kit
20 mL
None
10mL formalin
Non-sterile
Screw Cap
8 vials
30/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions