missing translation for 'onlineSavingsMsg'
Learn More

Prostaglandin E2 98.0+%, TCI America™

Catalog No. P18841MG
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18841MG 1 mg
P188410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18841MG Supplier TCI America Supplier No. P18841MG
Only null left

Chemical Identifiers

CAS 363-24-6
Molecular Formula C20H32O5
Molecular Weight (g/mol) 352.471
MDL Number MFCD00077861
InChI Key XEYBRNLFEZDVAW-ARSRFYASSA-N
Synonym prostaglandin e2, dinoprostone, pge2, prostin e2, prepidil, cervidil, propess, minprositin e2, minprostin e2, dinoproston
PubChem CID 5280360
ChEBI CHEBI:15551
IUPAC Name (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid
SMILES CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 1 mg
Formula Weight 352.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Prostaglandin E2
RTECSNumber UK8000000
TSCA No
Recommended Storage Freezer (-20 °C)