Learn More
DuPont™ ProShield™ 10 Frock
Provides general protection against non-hazardous light liquid splashes and dry particulates.
Supplier: DuPont™ PB271SWH2X003000
Description
- Frock provides body-length jacket design with front zipper closure
- Mandarin collar
- Extends below hip
- Elastic wrists
- Serged seams
- Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ASTM™ D1117 standards.
Specifications
| 2X-Large | |
| White | |
| Zipper | |
| Serged | |
| Coveralls, elastic wrists |
| ProShield | |
| Single-use | |
| Elastic | |
| General maintenance, janitorial/cleaning and other dirty work |