missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Frock
Description
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles. Applications include: general maintenance, janitorial/cleaning and other dirty work.
- Frock provides body-length jacket design with front zipper closure
- Mandarin collar
- Extends below hip
- Elastic wrists
- Serged seams
- Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
| Size | X-Large |
| Material | ProShield |
| Color | White |
| Disposable | Single-use |
| Closure Type | Zipper |
| Cuff Style | Elastic |
| No. of Pockets | None |
| Length (Metric) | 106 cm |
| Length (English) | 41.75 in. |
| Product Type | General Purpose Frock |
| Show More |