missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Frock
Description
- Frock provides body-length jacket design with front zipper closure
- Mandarin collar
- Extends below hip
- Elastic wrists
- Serged seams
- Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
| Size | Large |
| Material | ProShield |
| Color | White |
| Closure Type | Zipper |
| Cuff Style | Elastic |
| Seam Type | Serged |
| For Use With (Application) | General Protection |
| No. of Pockets | None |
| Length (Metric) | 40.75 |