missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Frock

Catalog No. 19162614
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Color:
Blue
White
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19162614 Medium Blue
19162615 X-Large Blue
19162610 2X-Large Blue
19162617 Medium White
19162616 Large White
19162618 X-Large White
19162653 2X-Large White
19162654 3X-Large White
19162655 4X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19162614 Supplier DuPont™ Supplier No. PB271SBUMD003000

Provides general protection against non-hazardous light liquid splashes and dry particulates.

  • Frock provides body-length jacket design with front zipper closure
  • Mandarin collar
  • Extends below hip
  • Elastic wrists
  • Serged seams
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Size Medium
Material ProShield
Color Blue
Closure Type Zipper
Cuff Style Elastic
No. of Pockets None
Length (Metric) 39.75
Seam Type Serged
For Use With (Application) General Protection