missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Frock

Catalog No. 19162610
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Color:
Blue
White
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19162610 2X-Large Blue
19162614 Medium Blue
19162615 X-Large Blue
19162617 Medium White
19162616 Large White
19162618 X-Large White
19162653 2X-Large White
19162654 3X-Large White
19162655 4X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19162610 Supplier DuPont™ Supplier No. PB271SBU2X003000
Only null left

Provides general protection against non-hazardous light liquid splashes and dry particulates.

  • Frock provides body-length jacket design with front zipper closure
  • Mandarin collar
  • Extends below hip
  • Elastic wrists
  • Serged seams
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Size 2X-Large
Material ProShield
Color Blue
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
For Use With (Application) General Protection
No. of Pockets None
Length (Metric) 42.5