Learn More
DuPont™ ProShield™ 6 SFR Hooded Coverall
6 SFR protective coveralls with standard fit hood and attached socks help extend the life of your flame resistant garments.
Supplier:  DuPont™ TM122SBUMD002500
 View more versions of this product
Catalog No. 17-500-169
Only null left
Explore available promotions
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps
Specifications
Specifications
Medium
Proshield
Single-use
Elastic
Zipper
Non-sterile
None
Unisex
Blue
Attached Socks, Elastic
Hooded
Serged
Flame, Particle
25/Cs.
Product Suggestions
Product Suggestions