Learn More
DuPont™ ProShield™ 6 SFR Hooded Coverall
Catalog No. 17500160
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500160 2X-Large
17500162 3X-Large
17500164 4X-Large
17500166 5X-Large
17500167 Large
17500169 Medium
17500171 X-Large
19165317 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17500160 Supplier DuPont™ Supplier No. TM122SBU2X002500
6 SFR protective coveralls with standard fit hood and attached socks help extend the life of your flame resistant garments.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps
Specifications
Size 2X-Large
Gender Unisex
Material Proshield
Color Blue
Disposable Single-use
Ankle Style Attached Socks, Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Flame, Particle
No. of Pockets None
Quantity 25/Cs.
Show More Show Less