missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 50 Coveralls with Hood

Catalog No. 19065503G
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Size (English) Chest:
29.75 to 33.25 in.
33.25 to 37.25 in.
37.25 to 40.75 in.
40.75 to 44.25
44.25 to 47.75 in.
47.75 to 51.25 in.
51.25 to 54.75 in.
54.75 to 58.25 in.
57.25 to 60.75 in.
Height (English):
5 to 5.58 ft.
5.22 to 6.16 ft.
5.25 to 5.58 ft.
5.41 to 5.75 ft.
6 to 6.33 ft.
6.16 to 6.33 ft.
6.33 to 6.58 ft.
6.58 to 6.83 ft.
6.75 to 7.08 ft.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest Height (English)
19065503G 4X-Large 51.25 to 54.75 in. 6.33 to 6.58 ft.
19065503A Small 29.75 to 33.25 in. 5 to 5.58 ft.
19065503B Medium 33.25 to 37.25 in. 5.25 to 5.58 ft.
19065503C Large 37.25 to 40.75 in. 5.41 to 5.75 ft.
19065503D X-Large 40.75 to 44.25 5.22 to 6.16 ft.
19065503E 2X-Large 44.25 to 47.75 in. 6 to 6.33 ft.
19065503F 3X-Large 47.75 to 51.25 in. 6.16 to 6.33 ft.
19065503H 5X-Large 54.75 to 58.25 in. 6.58 to 6.83 ft.
19065503I 6X-Large 57.25 to 60.75 in. 6.75 to 7.08 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Catalog No. 19065503G Supplier DuPont™ Supplier No. NB127SWH4X002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Should not be used around heat, flame, sparks or potentially flammable or explosive environments.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

  • Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.
  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Front zipper closure
  • Elastic wrists, ankles
  • Attached hood
  • Serged seams
  • Collar
  • White
  • Thickness: 10 mil

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Non-toxic Particulates
Model NB127S
Size (English) Chest 51.25 to 54.75 in.
Size (English) Inseam 29.25 in.
Size (Metric) Inseam 74.29 cm
Thickness 10 mil
For Use With (Application) Janitorial, Sanitation, General Industrial, Maintenance
Length (English) Sleeve 37.5 in.
Length (Metric) Sleeve 95.25 cm
Width (English) Chest 32.25 in.
Width (Metric) Chest 81.91 cm
Height (English) 6.33 to 6.58 ft.
Height (Metric) 1.93 to 2.01 m
Quantity 25/Cs.
Show More Show Less