missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 50 Coverall (all)

Catalog No. 19065502A
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
6X-Large
Height (English):
5 to 5.58 ft.
5.22 to 6.16 ft.
5.25 to 5.58 ft.
5.41 to 5.75 ft.
6 to 6.33 ft.
6.16 to 6.33 ft.
6.58 to 6.83 ft.
6.75 to 7.08 ft.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
19065502A Small 5 to 5.58 ft.
19065502B Medium 5.25 to 5.58 ft.
19065502C Large 5.41 to 5.75 ft.
19065502D X-Large 5.22 to 6.16 ft.
19065502E 2X-Large 6 to 6.33 ft.
19065502F 3X-Large 6.16 to 6.33 ft.
19065502H 5X-Large 6.58 to 6.83 ft.
19065502I 6X-Large 6.75 to 7.08 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Catalog No. 19065502A Supplier DuPont™ Supplier No. NB125SWHSM002500
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type Single-use Coveralls
Size Small
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Model NB125S
Size (English) Chest 29.75 to 33.25 in.
Size (English) Inseam 26.5 in.
Size (Metric) Inseam 67.31 cm
Thickness 10 mil
For Use With (Application) Janitorial, Sanitation, General Industrial, Maintenance
Length (English) Sleeve 31.25 in.
Length (Metric) Sleeve 79.38 cm
Width (English) Chest 21.5 in.
Width (Metric) Chest 54.61 cm
Height (English) 5 to 5.58 ft.
Height (Metric) 1.52 to 1.70 m
Quantity 25/Cs.
Show More Show Less